missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenylglyoxal hydrate, 95%, dry wt. basis

Catalog No. aal2027406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2027406 5 g
AAL2027403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2027406 Supplier Thermo Scientific Chemicals Supplier No. L2027406
Only null left

CAS: 16208-17-6 | C9H10O4 | 182.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16208-17-6
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00234534
InChI Key NLUFPYPVEHQAQN-UHFFFAOYSA-N
Synonym 4-methoxyphenylglyoxal hydrate, 2-4-methoxyphenyl-2-oxoacetaldehyde hydrate, pubchem12129, acmc-20ai17, 4-methoxyphenyl glyoxal hydrate, p-methoxyphenylglyoxal monohydrate, 2-4-methoxyphenyl-2-oxoethanal, hydrate, ethanone,2,2-dihydroxy-1-4-methoxyphenyl, 4-methoxyphenyl oxo acetaldehyde-water 1/1, 2-4-methoxyphenyl-2-oxidanylidene-ethanal hydrate
PubChem CID 15556730
IUPAC Name 2-(4-methoxyphenyl)-2-oxoacetaldehyde;hydrate
SMILES COC1=CC=C(C=C1)C(=O)C(O)O

Specifications

Melting Point 104°C to 111°C
Boiling Point 145°C to 147°C (13 mmHg)
Quantity 5 g
Beilstein 606701
Formula Weight 164.16 (Anhydrous)
Percent Purity 95%
Assay (dry wt.)
Chemical Name or Material 4-Methoxyphenylglyoxal hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only