missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenylacetone, 97+%

Catalog No. aaa1320922
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1320922 100 g
AAA1320914 25 g
AAA1320936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1320922 Supplier Thermo Scientific Chemicals Supplier No. A1320922
Only null left

CAS: 122-84-9 | C10H12O2 | 164.204 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 122-84-9
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00008773
InChI Key WFWKNGZODAOLEO-UHFFFAOYSA-N
Synonym 4-methoxyphenylacetone, p-acetonylanisole, anisketone, 1-4-methoxyphenyl propan-2-one, 2-propanone, 1-4-methoxyphenyl, anisyl methyl ketone, p-methoxyphenylacetone, 4-methoxybenzyl methyl ketone, 1-p-methoxyphenyl-2-propanone, anisic ketone
PubChem CID 31231
IUPAC Name 1-(4-methoxyphenyl)propan-2-one
SMILES CC(=O)CC1=CC=C(C=C1)OC

Specifications

Density 1.069
Boiling Point 266°C to 268°C
Flash Point 101°C (213°F)
Refractive Index 1.525
Quantity 100 g
Beilstein 2044332
Formula Weight 164.2
Percent Purity 98%
Chemical Name or Material 4-Methoxyphenylacetone
EINECSNumber 204-578-7
RTECSNumber UC2990000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only