accessibility menu, dialog, popup

UserName

H3COCH3O

CAS RN 122-84-9

IUPAC Name: 1-(4-methoxyphenyl)propan-2-one
Synonyms: P-anisyl methyl ketone
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: WFWKNGZODAOLEO-UHFFFAOYSA-N
SMILES: COC1=CC=C(CC(C)=O)C=C1
Boiling Point 
  • (6)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (6)
  • (2)
  • (3)
  • (5)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)