missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxyphenylacetic Acid 98.0+%, TCI America™
Supplier: TCI America M0742100G
Specifications
| 4-Methoxyphenylacetic Acid | |
| 86°C | |
| 305°C | |
| MFCD00004345 | |
| 4-methoxyphenylacetic acid, homoanisic acid, 2-4-methoxyphenyl acetic acid, 4-methoxybenzeneacetic acid, benzeneacetic acid, 4-methoxy, 4-methoxyphenyl acetic acid, p-methoxyphenylacetic acid, 2-p-anisyl acetic acid, p-methoxyphenyl acetic acid | |
| COC1=CC=C(C=C1)CC(=O)O | |
| 166.176 | |
| CHEBI:55501 | |
| ≥98.0% (GC,T) |
| 104-01-8 | |
| White-Yellow | |
| C9H10O3 | |
| 100 g | |
| NRPFNQUDKRYCNX-UHFFFAOYSA-N | |
| 2-(4-methoxyphenyl)acetic acid | |
| 7690 | |
| 166.18 | |
| Crystalline Powder |
Chemical Identifiers
| 104-01-8 | |
| 166.176 | |
| NRPFNQUDKRYCNX-UHFFFAOYSA-N | |
| 7690 | |
| 2-(4-methoxyphenyl)acetic acid |
| C9H10O3 | |
| MFCD00004345 | |
| 4-methoxyphenylacetic acid, homoanisic acid, 2-4-methoxyphenyl acetic acid, 4-methoxybenzeneacetic acid, benzeneacetic acid, 4-methoxy, 4-methoxyphenyl acetic acid, p-methoxyphenylacetic acid, 2-p-anisyl acetic acid, p-methoxyphenyl acetic acid | |
| CHEBI:55501 | |
| COC1=CC=C(C=C1)CC(=O)O |
Safety and Handling
EINECSNumber : (3)-3518
RTECSNumber : AI8960000
TSCA : Yes