missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenylacetic Acid 98.0+%, TCI America™

Catalog No. p-7132336
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Catalog No. Quantity
M074225G 25 g
M0742100G 100 g
M0742500G 500 g
3 options

Catalog No. M074225G

Supplier: TCI America M074225G

Only null left

Chemical Identifiers

CAS 104-01-8
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00004345
InChI Key NRPFNQUDKRYCNX-UHFFFAOYSA-N
Synonym 4-methoxyphenylacetic acid, homoanisic acid, 2-4-methoxyphenyl acetic acid, 4-methoxybenzeneacetic acid, benzeneacetic acid, 4-methoxy, 4-methoxyphenyl acetic acid, p-methoxyphenylacetic acid, 2-p-anisyl acetic acid, p-methoxyphenyl acetic acid
PubChem CID 7690
ChEBI CHEBI:55501
IUPAC Name 2-(4-methoxyphenyl)acetic acid
SMILES COC1=CC=C(C=C1)CC(=O)O

Specifications

Melting Point 86°C
Color White-Yellow
Boiling Point 305°C
Quantity 25 g
Formula Weight 166.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxyphenylacetic Acid
EINECSNumber (3)-3518
RTECSNumber AI8960000
TSCA Yes