missing translation for 'onlineSavingsMsg'
Learn More

(4-Methoxyphenyl)thiourea 98.0+%, TCI America™

Catalog No. p-7133312
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25935G 5 g
M259325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M25935G Supplier TCI America Supplier No. M25935G
Only null left

Chemical Identifiers

CAS 2293-07-4
Molecular Formula C8H10N2OS
Molecular Weight (g/mol) 182.24
MDL Number MFCD00004936
InChI Key SRYLJBWDZZMDSK-UHFFFAOYSA-N
Synonym 1-4-methoxyphenyl thiourea, 4-methoxyphenyl thiourea, n-4-methoxyphenyl thiourea, 1-4-methoxyphenyl-2-thiourea, p-anisylthiourea, n-p-anisylthiourea, p-methoxyphenyl thiourea, thiourea, 4-methoxyphenyl, n-p-methoxyphenyl thiourea
PubChem CID 667549
IUPAC Name (4-methoxyphenyl)thiourea
SMILES COC1=CC=C(C=C1)NC(=S)N

Specifications

Melting Point 208°C
Quantity 5 g
UN Number 2811
Formula Weight 182.24
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material (4-Methoxyphenyl)thiourea
RTECSNumber YT6825000
TSCA No