accessibility menu, dialog, popup

UserName

CAS RN 2293-07-4

IUPAC Name: (4-methoxyphenyl)thiourea
Synonyms: 4-methoxyphenylthiourea
Molecular Weight (g/mol): 182.24
Molecular Formula: C8H10N2OS
InChi Key: SRYLJBWDZZMDSK-UHFFFAOYSA-N
SMILES: COC1=CC=C(NC(N)=S)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results