missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenyl isothiocyanate, 98%

Catalog No. p-7022512
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1533104 2 g
AAA1533109 10 g
AAA1533118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1533104 Supplier Thermo Scientific Chemicals Supplier No. A1533104
Only null left

CAS: 2284-20-0 | C8H7NOS | 165.21 g/mol

4-Methoxyphenyl isothiocyanate has been used in the synthesis of 3-benzyl-2-(4-methoxyphenyl)-3H-[1,2,4]triazolo-[5,1-b]quinazolin-9-one and 2-{[4-amino-3-(4-methylphenyl)-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]acetyl}-N-arylhydrazinecarbothioamides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxyphenyl isothiocyanate has been used in the synthesis of 3-benzyl-2-(4-methoxyphenyl)-3H-[1,2,4]triazolo-[5,1-b]quinazolin-9-one and 2-{[4-amino-3-(4-methylphenyl)-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]acetyl}-N-arylhydrazinecarbothioamides.

Solubility
It hydrolyzes in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents, alcohols and amines.

Chemical Identifiers

CAS 2284-20-0
Molecular Formula C8H7NOS
Molecular Weight (g/mol) 165.21
MDL Number MFCD00011676
InChI Key VRPQCVLBOZOYCG-UHFFFAOYSA-N
Synonym 4-methoxyphenyl isothiocyanate, 4-methoxyphenylisothiocyanate, 1-isothiocyanato-4-methoxy-benzene, p-methoxyphenyl isothiocyanate, benzene, 1-isothiocyanato-4-methoxy, isothiocyanic acid 4-methoxyphenyl ester, 4-methoxybenzenisothiocyanate, 4-isothiocyanatoanisole, acmc-1ccqx, p-methoxyphenylisothiocyanate
PubChem CID 75293
IUPAC Name 1-isothiocyanato-4-methoxybenzene
SMILES COC1=CC=C(C=C1)N=C=S

Specifications

Melting Point 18°C to 20°C
Density 1.196
Boiling Point 279°C to 281°C
Flash Point >110°C (230°F)
Odor Pungent
Refractive Index 1.6489
Quantity 2 g
UN Number UN2922
Beilstein 606967
Sensitivity Moisture sensitive
Solubility Information It hydrolyzes in water.
Formula Weight 165.22
Percent Purity 98%
Chemical Name or Material 4-Methoxyphenyl isothiocyanate
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 218-921-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only