accessibility menu, dialog, popup

UserName

CAS RN 2284-20-0

IUPAC Name: 1-isothiocyanato-4-methoxybenzene
Synonyms: 1-isothiocyanato-4-methoxybenzene
Molecular Weight (g/mol): 165.21
Molecular Formula: C8H7NOS
InChi Key: VRPQCVLBOZOYCG-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)N=C=S
Boiling Point 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)