missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxyphenyl 3-O-Allyl-6-O-benzyl-2-deoxy-2-phthalimido-beta-D-glucopyranoside 98.0+%, TCI America™
$159.49 - $586.67
Chemical Identifiers
| CAS | 1820583-64-9 |
|---|---|
| Molecular Formula | C31H31NO8 |
| Molecular Weight (g/mol) | 545.59 |
| MDL Number | MFCD16295191 |
| InChI Key | UGQLXVORDHWCEW-UHFFFAOYNA-N |
| PubChem CID | 91659133 |
| IUPAC Name | 2-{6-[(benzyloxy)methyl]-5-hydroxy-2-(4-methoxyphenoxy)-4-(prop-2-en-1-yloxy)oxan-3-yl}-2,3-dihydro-1H-isoindole-1,3-dione |
| SMILES | COC1=CC=C(OC2OC(COCC3=CC=CC=C3)C(O)C(OCC=C)C2N2C(=O)C3=CC=CC=C3C2=O)C=C1 |
Chemical Identifiers
| 1820583-64-9 | |
| 545.59 | |
| UGQLXVORDHWCEW-UHFFFAOYNA-N | |
| 2-{6-[(benzyloxy)methyl]-5-hydroxy-2-(4-methoxyphenoxy)-4-(prop-2-en-1-yloxy)oxan-3-yl}-2,3-dihydro-1H-isoindole-1,3-dione |
| C31H31NO8 | |
| MFCD16295191 | |
| 91659133 | |
| COC1=CC=C(OC2OC(COCC3=CC=CC=C3)C(O)C(OCC=C)C2N2C(=O)C3=CC=CC=C3C2=O)C=C1 |
Specifications
| 1820583-64-9 | |
| C31H31NO8 | |
| UGQLXVORDHWCEW-UHFFFAOYNA-N | |
| 2-{6-[(benzyloxy)methyl]-5-hydroxy-2-(4-methoxyphenoxy)-4-(prop-2-en-1-yloxy)oxan-3-yl}-2,3-dihydro-1H-isoindole-1,3-dione | |
| 91659133 | |
| ≥98.0% (HPLC) | |
| 4-Methoxyphenyl 3-O-Allyl-6-O-benzyl-2-deoxy-2-phthalimido-beta-D-glucopyranoside |
| White | |
| MFCD16295191 | |
| COC1=CC=C(OC2OC(COCC3=CC=CC=C3)C(O)C(OCC=C)C2N2C(=O)C3=CC=CC=C3C2=O)C=C1 | |
| 545.59 | |
| 545.59 | |
| Crystalline Powder |
Safety and Handling
TSCA : No