accessibility menu, dialog, popup

UserName

CAS RN 1820583-64-9

IUPAC Name: 2-{6-[(benzyloxy)methyl]-5-hydroxy-2-(4-methoxyphenoxy)-4-(prop-2-en-1-yloxy)oxan-3-yl}-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: 2-{6-[(benzyloxy)methyl]-5-hydroxy-2-(4-methoxyphenoxy)-4-(prop-2-en-1-yloxy)oxan-3-yl}isoindole-1,3-dione
Molecular Weight (g/mol): 545.59
Molecular Formula: C31H31NO8
InChi Key: UGQLXVORDHWCEW-UHFFFAOYNA-N
SMILES: COC1=CC=C(OC2OC(COCC3=CC=CC=C3)C(O)C(OCC=C)C2N2C(=O)C3=CC=CC=C3C2=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results