missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenol, 99%

Catalog No. AC126005000
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
25 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC126005000 500 g
AC126001000 100 g
AC126000010 1 kg
AC126000051 5 kg
AC126000250 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AC126005000 Supplier Thermo Scientific Chemicals Supplier No. 126005000

CAS: 150-76-5 | C7H8O2 | 124.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 150-76-5 , 123-31-9
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.14
MDL Number MFCD00002332
InChI Key NWVVVBRKAWDGAB-UHFFFAOYSA-N
Synonym mequinol, 4-hydroxyanisole, p-hydroxyanisole, p-methoxyphenol, hydroquinone monomethyl ether, phenol, 4-methoxy, leucobasal, mehq, leucodine b, mechinolum
PubChem CID 9015
ChEBI CHEBI:69441
IUPAC Name 4-methoxyphenol
SMILES COC1=CC=C(O)C=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 56.0°C to 57.0°C
Color Colorless to Yellow
Boiling Point 243.0°C
Flash Point 133°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Quantity 500 g
Beilstein 06,843
Solubility Information Solubility in water: 40g/L (25°C). Other solubilities: readily soluble in ethyl acetate, 455g/100g ethyl alcohol - 426g/100g acetone (25°C), 70g/100g benzene - 3g/100g carbon tetrachloride, (25°C)
Viscosity 8.5 mPa.s (65°C)
Formula Weight 124.14
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material 4-Methoxyphenol, Stabilized
Show More Show Less
missing translation for 'healthHazard1' GHS Signal Word: Warning
missing translation for 'healthHazard2' GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
Causes serious eye irritation.
Suspected of damaging the unborn child.
Harmful to aquatic life with long lasting effects.
missing translation for 'healthHazard3' GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Obtain special instructions before use.
If eye i
EINECSNumber 205-769-8

RUO – Research Use Only