missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenol, 99%, Spectrum™ Chemical

Catalog No. 18608263
Change view
Click to view available options
Quantity:
100 g
500 g
25 kg
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608263 100 g Amber Glass Bottle
18608265 500 g Amber Glass Bottle
18608264 25 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18608263 Supplier Spectrum Chemical Mfg Cor Supplier No. M2280100GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 150-76-5

Specifications

CAS Max % 1
Melting Point 54° to 57°C
Boiling Point 243 to 246°C
Packaging Amber Glass Bottle
Quantity 100 g
Percent Purity 0.99
Grade Reagent
Physical Form Crystalline Powder or Lumps

CAUTION: For use by qualified personnel for research and development or other authorized use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.