missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxybiphenyl 99.0+%, TCI America™
Supplier: TCI America M094425G
Specifications
| 4-Methoxybiphenyl | |
| 89°C | |
| C13H12O | |
| 25 g | |
| RHDYQUZYHZWTCI-UHFFFAOYSA-N | |
| 4-methoxy-1,1'-biphenyl | |
| 11943 | |
| ≥99.0% (GC) |
| 613-37-6 | |
| 157°C | |
| MFCD00014897 | |
| 4-methoxybiphenyl, 4-phenylanisole, p-methoxybiphenyl, 1,1'-biphenyl, 4-methoxy, 4-methoxy-1,1'-biphenyl, anisole, p-phenyl, p-phenylanisole, 4-methoxy-biphenyl, biphenyl, 4-methoxy, 1-methoxy-4-phenyl-benzene | |
| COC1=CC=C(C=C1)C1=CC=CC=C1 | |
| 184.24 | |
| 184.24 | |
| Crystalline Powder |
Chemical Identifiers
| 613-37-6 | |
| 184.24 | |
| RHDYQUZYHZWTCI-UHFFFAOYSA-N | |
| 11943 | |
| COC1=CC=C(C=C1)C1=CC=CC=C1 |
| C13H12O | |
| MFCD00014897 | |
| 4-methoxybiphenyl, 4-phenylanisole, p-methoxybiphenyl, 1,1'-biphenyl, 4-methoxy, 4-methoxy-1,1'-biphenyl, anisole, p-phenyl, p-phenylanisole, 4-methoxy-biphenyl, biphenyl, 4-methoxy, 1-methoxy-4-phenyl-benzene | |
| 4-methoxy-1,1'-biphenyl |
Safety and Handling
EINECSNumber : (4)-0825
RTECSNumber : BZ8850000
TSCA : Yes