missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxybiphenyl 99.0+%, TCI America™
$50.33 - $124.89
Chemical Identifiers
| CAS | 613-37-6 |
|---|---|
| Molecular Formula | C13H12O |
| Molecular Weight (g/mol) | 184.24 |
| MDL Number | MFCD00014897 |
| InChI Key | RHDYQUZYHZWTCI-UHFFFAOYSA-N |
| Synonym | 4-methoxybiphenyl, 4-phenylanisole, p-methoxybiphenyl, 1,1'-biphenyl, 4-methoxy, 4-methoxy-1,1'-biphenyl, anisole, p-phenyl, p-phenylanisole, 4-methoxy-biphenyl, biphenyl, 4-methoxy, 1-methoxy-4-phenyl-benzene |
| PubChem CID | 11943 |
| IUPAC Name | 4-methoxy-1,1'-biphenyl |
| SMILES | COC1=CC=C(C=C1)C1=CC=CC=C1 |
Chemical Identifiers
| 613-37-6 | |
| 184.24 | |
| RHDYQUZYHZWTCI-UHFFFAOYSA-N | |
| 11943 | |
| COC1=CC=C(C=C1)C1=CC=CC=C1 |
| C13H12O | |
| MFCD00014897 | |
| 4-methoxybiphenyl, 4-phenylanisole, p-methoxybiphenyl, 1,1'-biphenyl, 4-methoxy, 4-methoxy-1,1'-biphenyl, anisole, p-phenyl, p-phenylanisole, 4-methoxy-biphenyl, biphenyl, 4-methoxy, 1-methoxy-4-phenyl-benzene | |
| 4-methoxy-1,1'-biphenyl |
Specifications
| 613-37-6 | |
| 157°C | |
| MFCD00014897 | |
| 4-methoxybiphenyl, 4-phenylanisole, p-methoxybiphenyl, 1,1'-biphenyl, 4-methoxy, 4-methoxy-1,1'-biphenyl, anisole, p-phenyl, p-phenylanisole, 4-methoxy-biphenyl, biphenyl, 4-methoxy, 1-methoxy-4-phenyl-benzene | |
| COC1=CC=C(C=C1)C1=CC=CC=C1 | |
| 184.24 | |
| 184.24 | |
| Crystalline Powder |
| 89°C | |
| C13H12O | |
| 5 g | |
| RHDYQUZYHZWTCI-UHFFFAOYSA-N | |
| 4-methoxy-1,1'-biphenyl | |
| 11943 | |
| ≥99.0% (GC) | |
| 4-Methoxybiphenyl |
Safety and Handling
EINECSNumber : (4)-0825
RTECSNumber : BZ8850000
TSCA : Yes