missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzylchloride, 98%, stabilized

Catalog No. AC294171000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC294171000 100 g
AC294170050 5 g
AC294170250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC294171000 Supplier Thermo Scientific Chemicals Supplier No. 294171000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 824-94-2 | C8H9ClO | 156.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 824-94-2 , 584-08-7
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.61
MDL Number MFCD00000915
InChI Key MOHYOXXOKFQHDC-UHFFFAOYSA-N
Synonym 4-methoxybenzyl chloride, 4-methoxybenzylchloride, 1-chloromethyl-4-methoxybenzene, p-methoxybenzyl chloride, p-anisyl chloride, alpha-chloro-4-methoxytoluene, p-chloromethyl anisole, benzene, 1-chloromethyl-4-methoxy, 4-chloromethyl anisole, anisole, p-chloromethyl
PubChem CID 69993
IUPAC Name 1-(chloromethyl)-4-methoxybenzene
SMILES COC1=CC=C(C=C1)CCl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -1.0°C
Color Colorless to Yellow
Density 1.1500g/mL
Boiling Point 117.0°C to 118.0°C (14.0 mmHg)
Flash Point 109°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Refractive Index 1.5470 to 1.5490
Quantity 100 g
UN Number 3265
Specific Gravity 1.15
Solubility Information Solubility in water: slowly decomposes
Stabilizer 1% K2CO3
Formula Weight 156.61
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Methoxybenzylchloride, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 212-540-6

RUO – Research Use Only