missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzylamine, 98+%

CAS: 2393-23-9 | C8H11NO | 137.182 g/mol

$107.85 - $1504.04

Chemical Identifiers

CAS 2393-23-9
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.182
MDL Number MFCD00008122
InChI Key IDPURXSQCKYKIJ-UHFFFAOYSA-N
Synonym 4-methoxybenzylamine, 4-methoxyphenyl methanamine, benzenemethanamine, 4-methoxy, p-methoxybenzylamine, 4-methoxy-benzylamine, anisylamine, benzylamine, p-methoxy, 4-methoxybenzyl amine, 1-4-methoxyphenyl methanamine, 4-methoxyphenyl methylamine
PubChem CID 75452
ChEBI CHEBI:49837
IUPAC Name (4-methoxyphenyl)methanamine
SMILES COC1=CC=C(C=C1)CN
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1499914
View Documents
Thermo Scientific Chemicals
A1499914
25 g
Each for $107.85
Only null left
 
AAA1499922
View Documents
Thermo Scientific Chemicals
A1499922
100 g
Each for $372.48
Only null left
 
AAA1499936
View Documents
Thermo Scientific Chemicals
A1499936
500 g
Each for $1,504.04
Only null left
 
Description

Description

4-Methoxybenzylamine is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff. It is also used fine chemical intemediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxybenzylamine is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff. It is also used fine chemical intemediate.

Solubility
It is highly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Moisture sensitive. Store under inert gas. Keep away from Strong oxidizing agentsacids, Acid chlorides, Acid anhydrides, Oxidizing agents, Chloroformates.
Specifications

Chemical Identifiers

2393-23-9
137.182
IDPURXSQCKYKIJ-UHFFFAOYSA-N
75452
(4-methoxyphenyl)methanamine
C8H11NO
MFCD00008122
4-methoxybenzylamine, 4-methoxyphenyl methanamine, benzenemethanamine, 4-methoxy, p-methoxybenzylamine, 4-methoxy-benzylamine, anisylamine, benzylamine, p-methoxy, 4-methoxybenzyl amine, 1-4-methoxyphenyl methanamine, 4-methoxyphenyl methylamine
CHEBI:49837
COC1=CC=C(C=C1)CN

Specifications

2393-23-9
1.053
114°C (237°F)
C8H11NO
MFCD00008122
UN2735
Air sensitive; May form a precipitate
It is highly soluble in water.
COC1=CC=C(C=C1)CN
137.182
CHEBI:49837
≥98%
-10°C
236°C to 237°C
Amine-like
1.546
25 g
508206
4-methoxybenzylamine, 4-methoxyphenyl methanamine, benzenemethanamine, 4-methoxy, p-methoxybenzylamine, 4-methoxy-benzylamine, anisylamine, benzylamine, p-methoxy, 4-methoxybenzyl amine, 1-4-methoxyphenyl methanamine, 4-methoxyphenyl methylamine
IDPURXSQCKYKIJ-UHFFFAOYSA-N
(4-methoxyphenyl)methanamine
75452
137.18
4-Methoxybenzylamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

EINECSNumber : 219-247-2

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

RUO – Research Use Only