missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzylamine, 98+%

CAS: 2393-23-9 | C8H11NO | 137.182 g/mol

Supplier:  Thermo Scientific Chemicals A1499922

 View more versions of this product

Catalog No. AAA1499922


Only null left
Explore available promotions

Description

Description

4-Methoxybenzylamine is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff. It is also used fine chemical intemediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxybenzylamine is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff. It is also used fine chemical intemediate.

Solubility
It is highly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Moisture sensitive. Store under inert gas. Keep away from Strong oxidizing agentsacids, Acid chlorides, Acid anhydrides, Oxidizing agents, Chloroformates.
Specifications

Specifications

4-Methoxybenzylamine
-10°C
236°C to 237°C
Amine-like
1.546
100 g
508206
4-methoxybenzylamine, 4-methoxyphenyl methanamine, benzenemethanamine, 4-methoxy, p-methoxybenzylamine, 4-methoxy-benzylamine, anisylamine, benzylamine, p-methoxy, 4-methoxybenzyl amine, 1-4-methoxyphenyl methanamine, 4-methoxyphenyl methylamine
IDPURXSQCKYKIJ-UHFFFAOYSA-N
(4-methoxyphenyl)methanamine
75452
137.18
2393-23-9
1.053
114°C (237°F)
C8H11NO
MFCD00008122
UN2735
Air sensitive; May form a precipitate
It is highly soluble in water.
COC1=CC=C(C=C1)CN
137.182
CHEBI:49837
≥98%

Chemical Identifiers

2393-23-9
137.182
IDPURXSQCKYKIJ-UHFFFAOYSA-N
75452
(4-methoxyphenyl)methanamine
C8H11NO
MFCD00008122
4-methoxybenzylamine, 4-methoxyphenyl methanamine, benzenemethanamine, 4-methoxy, p-methoxybenzylamine, 4-methoxy-benzylamine, anisylamine, benzylamine, p-methoxy, 4-methoxybenzyl amine, 1-4-methoxyphenyl methanamine, 4-methoxyphenyl methylamine
CHEBI:49837
COC1=CC=C(C=C1)CN
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

EINECSNumber : 219-247-2

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only