missing translation for 'onlineSavingsMsg'
Learn More

(4-Methoxybenzyl) Methyl Ether 98.0+%, TCI America™

Catalog No. M23055G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M23055G Supplier TCI America Supplier No. M23055G

Chemical Identifiers

CAS 1515-81-7
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00155135
InChI Key RSOYRXBYZFBWFS-UHFFFAOYSA-N
Synonym 4-(Methoxymethyl)anisole
PubChem CID 73689
IUPAC Name 1-methoxy-4-(methoxymethyl)benzene
SMILES COCC1=CC=C(OC)C=C1

Specifications

Color Yellow
Boiling Point 226°C
Quantity 5 g
Formula Weight 152.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (4-Methoxybenzyl) Methyl Ether