accessibility menu, dialog, popup

UserName

CAS RN 1515-81-7

IUPAC Name: 1-methoxy-4-(methoxymethyl)benzene
Synonyms: 1-methoxy-4-(methoxymethyl)benzene
Molecular Weight (g/mol): 152.19
Molecular Formula: C9H12O2
InChi Key: RSOYRXBYZFBWFS-UHFFFAOYSA-N
SMILES: COCC1=CC=C(OC)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results