missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzyl isothiocyanate, 94%

Catalog No. p-7045880
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1234203 1 g
AAL1234206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1234203 Supplier Thermo Scientific Chemicals Supplier No. L1234203
Only null left

CAS: 3694-57-3 | C9H9NOS | 179.237 g/mol

4-Methoxybenzyl isothiocyanate is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxybenzyl isothiocyanate is used as pharmaceutical intermediates.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Incompatible with bases, alcohols and amines. Store under dry inert gas. Protect from humidity and water. Store away from moisture.

Chemical Identifiers

CAS 3694-57-3
Molecular Formula C9H9NOS
Molecular Weight (g/mol) 179.237
MDL Number MFCD00041117
InChI Key IMFQYAJJXFXVMM-UHFFFAOYSA-N
Synonym 4-methoxybenzyl isothiocyanate, 1-isothiocyanatomethyl-4-methoxybenzene, 4-methoxybenzylisothiocyanate, benzene, 1-isothiocyanatomethyl-4-methoxy, p-methoxybenzyl isothiocyanate, 1-isothiocyanatomethyl-4-methoxy-benzene, chembl81201, 4-methoxybenzyl isothiocyanate 5gr, acmc-1cq6l
PubChem CID 123197
IUPAC Name 1-(isothiocyanatomethyl)-4-methoxybenzene
SMILES COC1=CC=C(C=C1)CN=C=S

Specifications

Density 1.15
Boiling Point 132°C to 133°C (0.7 mmHg)
Odor Irritating
Refractive Index 1.5975
Quantity 1 g
UN Number UN2922
Beilstein 2089549
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 179.24
Percent Purity 94%
Chemical Name or Material 4-Methoxybenzyl isothiocyanate
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only