missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzyl alcohol, 98%

Catalog No. AC159731000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC159731000 100 g Glass bottle
AC159735000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC159731000 Supplier Thermo Scientific Chemicals Supplier No. 159731000
Only null left

CAS: 105-13-5 | C8H10O2 | 138.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 105-13-5
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00004653
InChI Key MSHFRERJPWKJFX-UHFFFAOYSA-N
Synonym 4-methoxybenzyl alcohol, 4-methoxyphenyl methanol, anise alcohol, anisyl alcohol, p-anisyl alcohol, p-methoxybenzyl alcohol, benzenemethanol, 4-methoxy, anisic alcohol, p-anisol alcohol, 4-methoxybenzenemethanol
PubChem CID 7738
ChEBI CHEBI:86918
IUPAC Name (4-methoxyphenyl)methanol
SMILES COC1=CC=C(CO)C=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 23.0°C to 25.5°C
Color Colorless to Yellow
Density 1.1125g/mL
Boiling Point 259.0°C
Flash Point 146°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5430 to 1.5460
Linear Formula CH3OC6H4CH2OH
Quantity 100 g
Beilstein 06,897
Merck Index 15,658
Specific Gravity 1.1125
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in 1 volume of 50% alcohol,freely soluble in ether
Formula Weight 138.17
Percent Purity 98%
Physical Form Liquid After Melting
Chemical Name or Material 4-Methoxybenzyl alcohol, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with wa
EINECSNumber 203-273-6
RTECSNumber DO8925000
TSCA TSCA

RUO – Research Use Only