missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzyl Acetate 98.0+%, TCI America™

Catalog No. A088625ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A088625ML 25 mL
A0886500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A088625ML Supplier TCI America Supplier No. A088625ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 104-21-2
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
MDL Number MFCD00038509
InChI Key HFNGYHHRRMSKEU-UHFFFAOYSA-N
Synonym 4-methoxybenzyl acetate, anisyl acetate, cassie ketone, 4-methoxyphenyl methyl acetate, p-anisyl acetate, p-methoxybenzyl acetate, p-methoxybenzyl alcohol acetate, benzyl alcohol, p-methoxy-, acetate, benzenemethanol, 4-methoxy-, acetate, benzenemethanol, 4-methoxy-, 1-acetate
PubChem CID 7695
ChEBI CHEBI:86743
IUPAC Name (4-methoxyphenyl)methyl acetate
SMILES COC1=CC=C(COC(C)=O)C=C1

Specifications

Color Yellow
Boiling Point 137°C
Quantity 25 mL
Formula Weight 180.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methoxybenzyl Acetate
EINECSNumber (3)-1057&(3)-1069
TSCA Yes