accessibility menu, dialog, popup

UserName

CAS RN 104-21-2

IUPAC Name: (4-methoxyphenyl)methyl acetate
Synonyms: anisyl acetate
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: HFNGYHHRRMSKEU-UHFFFAOYSA-N
SMILES: COC1=CC=C(COC(C)=O)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results