missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzhydrol 98.0+%, TCI America™

Catalog No. M12785G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12785G 5 g
M127825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M12785G Supplier TCI America Supplier No. M12785G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 720-44-5
Molecular Formula C14H14O2
Molecular Weight (g/mol) 214.264
MDL Number MFCD00014398
InChI Key BEGZWXVLBIZFKQ-UHFFFAOYSA-N
Synonym 4-methoxybenzhydrol, 4-methoxyphenyl phenyl methanol, 4-methoxybenzhydryl alcohol, benzenemethanol, 4-methoxy-.alpha.-phenyl, 4-methoxyphenyl phenylmethan-1-ol, 4-methoxy-phenyl-phenyl-methanol, mamp-oh resin, 4-methoxybenzohydrol, acmc-209olf, p-methoxybenzhydryl alcohol
PubChem CID 95375
IUPAC Name (4-methoxyphenyl)-phenylmethanol
SMILES COC1=CC=C(C=C1)C(C2=CC=CC=C2)O

Specifications

Melting Point 65°C
Color White-Yellow
Quantity 5 g
Formula Weight 214.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxybenzhydrol