accessibility menu, dialog, popup

UserName

CAS RN 720-44-5

IUPAC Name: (4-methoxyphenyl)(phenyl)methanol
Synonyms: (4-methoxyphenyl)(phenyl)methanol
Molecular Weight (g/mol): 214.26
Molecular Formula: C14H14O2
InChi Key: BEGZWXVLBIZFKQ-UHFFFAOYNA-N
SMILES: COC1=CC=C(C=C1)C(O)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)