missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzhydrol, 98+%

Catalog No. AAL0046806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0046806 5 g
AAL0046814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0046806 Supplier Thermo Scientific Chemicals Supplier No. L0046806

CAS: 720-44-5 | C14H14O2 | 214.264 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 720-44-5
Molecular Formula C14H14O2
Molecular Weight (g/mol) 214.264
MDL Number MFCD00014398
InChI Key BEGZWXVLBIZFKQ-UHFFFAOYSA-N
Synonym 4-methoxybenzhydrol, 4-methoxyphenyl phenyl methanol, 4-methoxybenzhydryl alcohol, benzenemethanol, 4-methoxy-.alpha.-phenyl, 4-methoxyphenyl phenylmethan-1-ol, 4-methoxy-phenyl-phenyl-methanol, mamp-oh resin, 4-methoxybenzohydrol, acmc-209olf, p-methoxybenzhydryl alcohol
PubChem CID 95375
IUPAC Name (4-methoxyphenyl)-phenylmethanol
SMILES COC1=CC=C(C=C1)C(C2=CC=CC=C2)O

Specifications

Melting Point 65°C to 68°C
Quantity 5 g
Beilstein 3202445
Formula Weight 214.26
Percent Purity ≥98%
Chemical Name or Material 4-Methoxybenzhydrol
EINECSNumber 211-953-9
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only