missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzaldehyde dimethyl acetal, 98%

Catalog No. p-7022848
Change view
Click to view available options
Quantity:
25 g
250 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1579314 25 g
AAA1579330 250 g
AAA1579336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1579314 Supplier Thermo Scientific Chemicals Supplier No. A1579314
Only null left

CAS: 2186-92-7 | C10H14O3 | 182.22 g/mol

4-Methoxybenzaldehyde dimethyl acetal is used as a precursor to 1-methoxy-1-(4-methoxyphenyl)hept-2-yne, 2,5-dioxopyrrolidin-1-yl-3-((4R)-2-(4-methoxyphenyl)-5,5-dimethyl-1,3-dioxane-4-carboxamido)propanoate and 1-(p-anisyl)-2-(2-methoxyethyl)-3-phenylindene. Further, it acts as a protecting group reagent for diols, especially in carbohydrates. It is also used as a flavor essence in sunflower, cyclamen and in jasmine. In addition to this, it is involved in allylation reactions with allyltrimethylsilane catalyzed by Iron(III) chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxybenzaldehyde dimethyl acetal is used as a precursor to 1-methoxy-1-(4-methoxyphenyl)hept-2-yne, 2,5-dioxopyrrolidin-1-yl-3-((4R)-2-(4-methoxyphenyl)-5,5-dimethyl-1,3-dioxane-4-carboxamido)propanoate and 1-(p-anisyl)-2-(2-methoxyethyl)-3-phenylindene. Further, it acts as a protecting group reagent for diols, especially in carbohydrates. It is also used as a flavor essence in sunflower, cyclamen and in jasmine. In addition to this, it is involved in allylation reactions with allyltrimethylsilane catalyzed by Iron(III) chloride.

Solubility
Miscible with alcohol and many organic solvents. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents, strong acids and strong bases. Store in a cool place.

Chemical Identifiers

CAS 2186-92-7
Molecular Formula C10H14O3
Molecular Weight (g/mol) 182.22
MDL Number MFCD00036507
InChI Key NNHYAHOTXLASEA-UHFFFAOYSA-N
Synonym 1-dimethoxymethyl-4-methoxybenzene, p-anisaldehyde dimethyl acetal, anisaldehyde dimethyl acetal, 4-methoxybenzaldehyde dimethyl acetal, p-dimethoxymethyl anisole, benzene, 1-dimethoxymethyl-4-methoxy, p-anisaldehydedimethylacetal, unii-xte77a5et4, anisicaldehyde dimethylacetal, p-anisaldehyde dimethylacetal
PubChem CID 75140
IUPAC Name 1-(dimethoxymethyl)-4-methoxybenzene
SMILES COC(OC)C1=CC=C(OC)C=C1

Specifications

Density 1.068
Boiling Point 253°C
Flash Point 97°C (206°F)
Odor Pleasant
Refractive Index 1.5055
Quantity 25 g
Beilstein 2048930
Solubility Information Miscible with alcohol and many organic solvents. Slightly miscible with water.
Formula Weight 182.22
Percent Purity 98%
Chemical Name or Material 4-Methoxybenzaldehyde dimethyl acetal
Show More Show Less
EINECSNumber 218-577-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only