missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-3-(trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Supplier:  TCI America M28391G

 View more versions of this product

Catalog No. M28391G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4-Methoxy-3-(trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride)
202°C
C8H8BF3O3
1 g
BUSMBMGODABSIN-UHFFFAOYSA-N
[4-methoxy-3-(trifluoromethyl)phenyl]boronic acid
17750052
Crystalline Powder
149507-36-8
White
MFCD07363790
4-methoxy-3-trifluoromethyl phenyl boronic acid, 4-methoxy-3-trifluoromethylphenylboronic acid, 3-trifluoromethyl-4-methoxy-phenylboronic acid, 4-methoxy-3-trifluoromethylphenyl boronic acid, 4-methoxy-3-trifluoromethyl benzeneboronic acid, 4-methoxy-3-trifluoromethyl phenylboronic acid, boronic acid, 4-methoxy-3-trifluoromethyl phenyl, acmc-1c5ik
B(C1=CC(=C(C=C1)OC)C(F)(F)F)(O)O
219.954
219.95

Chemical Identifiers

149507-36-8
219.954
BUSMBMGODABSIN-UHFFFAOYSA-N
17750052
B(C1=CC(=C(C=C1)OC)C(F)(F)F)(O)O
C8H8BF3O3
MFCD07363790
4-methoxy-3-trifluoromethyl phenyl boronic acid, 4-methoxy-3-trifluoromethylphenylboronic acid, 3-trifluoromethyl-4-methoxy-phenylboronic acid, 4-methoxy-3-trifluoromethylphenyl boronic acid, 4-methoxy-3-trifluoromethyl benzeneboronic acid, 4-methoxy-3-trifluoromethyl phenylboronic acid, boronic acid, 4-methoxy-3-trifluoromethyl phenyl, acmc-1c5ik
[4-methoxy-3-(trifluoromethyl)phenyl]boronic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions