accessibility menu, dialog, popup

UserName

CH3OFFFBOHHO

CAS RN 149507-36-8

IUPAC Name: [4-methoxy-3-(trifluoromethyl)phenyl]boronic acid
Synonyms: 4-methoxy-3-(trifluoromethyl)phenylboronic acid
Molecular Weight (g/mol): 219.95
Molecular Formula: C8H8BF3O3
InChi Key: BUSMBMGODABSIN-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1C(F)(F)F)B(O)O
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results