missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxy-3-nitropyridine 98.0+%, TCI America™
$161.72 - $469.74
Chemical Identifiers
| CAS | 31872-62-5 |
|---|---|
| Molecular Formula | C6H6N2O3 |
| Molecular Weight (g/mol) | 154.125 |
| MDL Number | MFCD00209661 |
| InChI Key | BZPVREXVOZITPF-UHFFFAOYSA-N |
| Synonym | 4-methoxy-3-nitro-pyridine, pyridine, 4-methoxy-3-nitro, methyl 3-nitro-4-pyridinyl ether, 3-nitro-4-methoxypyridine, methyl3-nitro-4-pyridinylether, pubchem14990, acmc-1ctwk, 3-nitro-4-methoxy pyridine, ksc494q6d, # |
| PubChem CID | 355832 |
| IUPAC Name | 4-methoxy-3-nitropyridine |
| SMILES | COC1=C(C=NC=C1)[N+](=O)[O-] |
Chemical Identifiers
| 31872-62-5 | |
| 154.125 | |
| BZPVREXVOZITPF-UHFFFAOYSA-N | |
| 355832 | |
| COC1=C(C=NC=C1)[N+](=O)[O-] |
| C6H6N2O3 | |
| MFCD00209661 | |
| 4-methoxy-3-nitro-pyridine, pyridine, 4-methoxy-3-nitro, methyl 3-nitro-4-pyridinyl ether, 3-nitro-4-methoxypyridine, methyl3-nitro-4-pyridinylether, pubchem14990, acmc-1ctwk, 3-nitro-4-methoxy pyridine, ksc494q6d, # | |
| 4-methoxy-3-nitropyridine |
Specifications
| 31872-62-5 | |
| Yellow | |
| C6H6N2O3 | |
| 1 g | |
| BZPVREXVOZITPF-UHFFFAOYSA-N | |
| 4-methoxy-3-nitropyridine | |
| 355832 | |
| ≥98.0% (GC) | |
| 4-Methoxy-3-nitropyridine |
| 76°C | |
| 127°C | |
| MFCD00209661 | |
| 4-methoxy-3-nitro-pyridine, pyridine, 4-methoxy-3-nitro, methyl 3-nitro-4-pyridinyl ether, 3-nitro-4-methoxypyridine, methyl3-nitro-4-pyridinylether, pubchem14990, acmc-1ctwk, 3-nitro-4-methoxy pyridine, ksc494q6d, # | |
| COC1=C(C=NC=C1)[N+](=O)[O-] | |
| 154.125 | |
| 154.13 | |
| Crystalline Powder |
Safety and Handling
TSCA : No