CAS RN 31872-62-5
CAS RN 31872-62-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Methoxy-3-nitropyridine 98.0+%, TCI America™
CAS: 31872-62-5 Molecular Formula: C6H6N2O3 Molecular Weight (g/mol): 154.125 MDL Number: MFCD00209661 InChI Key: BZPVREXVOZITPF-UHFFFAOYSA-N Synonym: 4-methoxy-3-nitro-pyridine,pyridine, 4-methoxy-3-nitro,methyl 3-nitro-4-pyridinyl ether,3-nitro-4-methoxypyridine,methyl3-nitro-4-pyridinylether,pubchem14990,acmc-1ctwk,3-nitro-4-methoxy pyridine,ksc494q6d,# PubChem CID: 355832 IUPAC Name: 4-methoxy-3-nitropyridine SMILES: COC1=C(C=NC=C1)[N+](=O)[O-]