missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-2-(trifluoromethyl)benzeneboronic acid

Catalog No. AAH2706203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2706203 1 g
AAH2706206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2706203 Supplier Thermo Scientific Chemicals Supplier No. H2706203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 313546-16-6 | C8H8BF3O3 | 219.954 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 313546-16-6
Molecular Formula C8H8BF3O3
Molecular Weight (g/mol) 219.954
MDL Number MFCD03095346
InChI Key ZBCRZEJNAADYKG-UHFFFAOYSA-N
Synonym 4-methoxy-2-trifluoromethyl phenylboronic acid, 4-methoxy-2-trifluoromethyl benzeneboronic acid, 4-methoxy-2-trifluoromethyl phenyl boronic acid, 4-methoxy-2-trifluoromethyl-phenylboronic acid, 4-methoxy-2, pubchem1868, acmc-1aefl, ablock ab-13-0076
PubChem CID 12109460
IUPAC Name [4-methoxy-2-(trifluoromethyl)phenyl]boronic acid
SMILES B(C1=C(C=C(C=C1)OC)C(F)(F)F)(O)O

Specifications

Melting Point 168°C to 172°C
Quantity 1 g
Formula Weight 219.95
Chemical Name or Material 4-Methoxy-2-(trifluoromethyl)benzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only