CAS RN 313546-16-6
IUPAC Name:
[4-methoxy-2-(trifluoromethyl)phenyl]boronic acid
Synonyms:
4-methoxy-2-(trifluoromethyl)phenylboronic acid
Molecular Weight (g/mol):
219.95
Molecular Formula:
C8H8BF3O3
InChi Key:
ZBCRZEJNAADYKG-UHFFFAOYSA-N
SMILES:
COC1=CC(=C(C=C1)B(O)O)C(F)(F)F
1
–
1
of
1
results
1
Promotions Available
4-Methoxy-2-(trifluoromethyl)benzeneboronic acid
4-Methoxy-2-(trifluoromethyl)benzeneboronic acid
CAS: 313546-16-6 Molecular Formula: C8H8BF3O3 Molecular Weight (g/mol): 219.954 MDL Number: MFCD03095346 InChI Key: ZBCRZEJNAADYKG-UHFFFAOYSA-N Synonym: 4-methoxy-2-trifluoromethyl phenylboronic acid,4-methoxy-2-trifluoromethyl benzeneboronic acid,4-methoxy-2-trifluoromethyl phenyl boronic acid,4-methoxy-2-trifluoromethyl-phenylboronic acid,4-methoxy-2,pubchem1868,acmc-1aefl,ablock ab-13-0076 PubChem CID: 12109460 IUPAC Name: [4-methoxy-2-(trifluoromethyl)phenyl]boronic acid SMILES: B(C1=C(C=C(C=C1)OC)C(F)(F)F)(O)O