missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxy-2-(trifluoromethyl)benzaldehyde 97.0+%, TCI America™
$109.74 - $109.74
Chemical Identifiers
| CAS | 106312-36-1 |
|---|---|
| Molecular Formula | C9H7F3O2 |
| Molecular Weight (g/mol) | 204.148 |
| MDL Number | MFCD01091011 |
| InChI Key | BVPVUMRIGHMFNV-UHFFFAOYSA-N |
| Synonym | 4-methoxy-2-trifluoromethyl benzaldehyde, benzaldehyde, 4-methoxy-2-trifluoromethyl, 4-formyl-3-trifluoromethyl anisole, pubchem1463, acmc-1c5gx, ksc504i7t, 2-formyl-5-methoxybenzotrifluoride, 4-methoxy-2-trifluoromethyl-benzaldehyde, benzaldehyde,4-methoxy-2-trifluoromethyl |
| PubChem CID | 2778286 |
| IUPAC Name | 4-methoxy-2-(trifluoromethyl)benzaldehyde |
| SMILES | COC1=CC(=C(C=C1)C=O)C(F)(F)F |
Chemical Identifiers
| 106312-36-1 | |
| 204.148 | |
| BVPVUMRIGHMFNV-UHFFFAOYSA-N | |
| 2778286 | |
| COC1=CC(=C(C=C1)C=O)C(F)(F)F |
| C9H7F3O2 | |
| MFCD01091011 | |
| 4-methoxy-2-trifluoromethyl benzaldehyde, benzaldehyde, 4-methoxy-2-trifluoromethyl, 4-formyl-3-trifluoromethyl anisole, pubchem1463, acmc-1c5gx, ksc504i7t, 2-formyl-5-methoxybenzotrifluoride, 4-methoxy-2-trifluoromethyl-benzaldehyde, benzaldehyde,4-methoxy-2-trifluoromethyl | |
| 4-methoxy-2-(trifluoromethyl)benzaldehyde |
Specifications
| 106312-36-1 | |
| White | |
| MFCD01091011 | |
| 4-methoxy-2-trifluoromethyl benzaldehyde, benzaldehyde, 4-methoxy-2-trifluoromethyl, 4-formyl-3-trifluoromethyl anisole, pubchem1463, acmc-1c5gx, ksc504i7t, 2-formyl-5-methoxybenzotrifluoride, 4-methoxy-2-trifluoromethyl-benzaldehyde, benzaldehyde,4-methoxy-2-trifluoromethyl | |
| COC1=CC(=C(C=C1)C=O)C(F)(F)F | |
| 204.148 | |
| 204.15 | |
| Crystalline Lumps |
| 40°C | |
| C9H7F3O2 | |
| 1 g | |
| BVPVUMRIGHMFNV-UHFFFAOYSA-N | |
| 4-methoxy-2-(trifluoromethyl)benzaldehyde | |
| 2778286 | |
| ≥97.0% (GC) | |
| 4-Methoxy-2-(trifluoromethyl)benzaldehyde |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator