missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-2-nitrobenzoic Acid 98.0+%, TCI America™

Catalog No. M28285G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28285G 5 g
M28281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M28285G Supplier TCI America Supplier No. M28285G

Chemical Identifiers

CAS 33844-21-2
Molecular Formula C8H7NO5
Molecular Weight (g/mol) 197.146
MDL Number MFCD04972109
InChI Key DVZBWONCSHFMMM-UHFFFAOYSA-N
Synonym 2-Nitro-p-anisic Acid
PubChem CID 288657
SMILES COC1=CC(=C(C=C1)C(=O)O)[N+](=O)[O-]

Specifications

Melting Point 200°C
Quantity 5 g
Formula Weight 197.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxy-2-nitrobenzoic Acid