accessibility menu, dialog, popup

UserName

H3COOHONOO

CAS RN 33844-21-2

IUPAC Name: 4-methoxy-2-nitrobenzoic acid
Synonyms: 4-methoxy-2-nitrobenzoic acid
Molecular Weight (g/mol): 197.15
Molecular Formula: C8H7NO5
InChi Key: DVZBWONCSHFMMM-UHFFFAOYSA-N
SMILES: COC1=CC=C(C(O)=O)C(=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results