missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxy-2-nitrobenzenesulfonyl Chloride 93.0+%, TCI America™
$100.67 - $264.70
Chemical Identifiers
| CAS | 18092-54-1 |
|---|---|
| Molecular Formula | C7H6ClNO5S |
| Molecular Weight (g/mol) | 251.637 |
| MDL Number | MFCD00143333 |
| InChI Key | OGHAPVZVXLHURO-UHFFFAOYSA-N |
| PubChem CID | 87452 |
| IUPAC Name | 4-methoxy-2-nitrobenzenesulfonyl chloride |
| SMILES | COC1=CC(=C(C=C1)S(=O)(=O)Cl)[N+](=O)[O-] |
Chemical Identifiers
| 18092-54-1 | |
| 251.637 | |
| OGHAPVZVXLHURO-UHFFFAOYSA-N | |
| 4-methoxy-2-nitrobenzenesulfonyl chloride |
| C7H6ClNO5S | |
| MFCD00143333 | |
| 87452 | |
| COC1=CC(=C(C=C1)S(=O)(=O)Cl)[N+](=O)[O-] |
Specifications
| 18092-54-1 | |
| Yellow | |
| MFCD00143333 | |
| 3261 | |
| COC1=CC(=C(C=C1)S(=O)(=O)Cl)[N+](=O)[O-] | |
| 251.637 | |
| 251.64 | |
| Crystalline Powder |
| 77°C | |
| C7H6ClNO5S | |
| 5 g | |
| OGHAPVZVXLHURO-UHFFFAOYSA-N | |
| 4-methoxy-2-nitrobenzenesulfonyl chloride | |
| 87452 | |
| ≥93.0% (T) | |
| 4-Methoxy-2-nitrobenzenesulfonyl Chloride |
Safety and Handling
TSCA : No