missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxy-2-nitrobenzenesulfonyl Chloride 93.0+%, TCI America™
Supplier: TCI America M100625G
Specifications
| 4-Methoxy-2-nitrobenzenesulfonyl Chloride | |
| 77°C | |
| C7H6ClNO5S | |
| 25 g | |
| OGHAPVZVXLHURO-UHFFFAOYSA-N | |
| 4-methoxy-2-nitrobenzenesulfonyl chloride | |
| 87452 | |
| ≥93.0% (T) |
| 18092-54-1 | |
| Yellow | |
| MFCD00143333 | |
| 3261 | |
| COC1=CC(=C(C=C1)S(=O)(=O)Cl)[N+](=O)[O-] | |
| 251.637 | |
| 251.64 | |
| Crystalline Powder |
Chemical Identifiers
| 18092-54-1 | |
| 251.637 | |
| OGHAPVZVXLHURO-UHFFFAOYSA-N | |
| 4-methoxy-2-nitrobenzenesulfonyl chloride |
| C7H6ClNO5S | |
| MFCD00143333 | |
| 87452 | |
| COC1=CC(=C(C=C1)S(=O)(=O)Cl)[N+](=O)[O-] |
Safety and Handling
TSCA : No