Learn More
4'-Methoxy-2'-nitroacetanilide, 98+%
CAS: 119-81-3 | C9H10N2O4 | 210.189 g/mol
Supplier: Thermo Scientific Chemicals L0421814
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 4'-Methoxy-2'-nitroacetanilide | |
| 117°C to 118°C | |
| MFCD00017018 | |
| 2809678 | |
| QGEGALJODPBPGR-UHFFFAOYSA-N | |
| N-(4-methoxy-2-nitrophenyl)acetamide | |
| 67075 | |
| ≥98% |
| 119-81-3 | |
| C9H10N2O4 | |
| 25 g | |
| n-4-methoxy-2-nitrophenyl acetamide, 4'-methoxy-2'-nitroacetanilide, 4-methoxy-2-nitroacetanilide, 2'-nitro-p-acetanisidide, acetamide, n-4-methoxy-2-nitrophenyl, 2-nitro-p-acetanisidide, p-acetanisidide, 2-nitro, 4-acetamido-3-nitroanisole, 2-nitro-4-methoxyacetanilide, p-acetanisidide, 2'-nitro | |
| CC(=O)NC1=C(C=C(C=C1)OC)[N+](=O)[O-] | |
| 210.189 | |
| 210.19 |
Chemical Identifiers
| 119-81-3 | |
| 210.189 | |
| QGEGALJODPBPGR-UHFFFAOYSA-N | |
| 67075 | |
| CC(=O)NC1=C(C=C(C=C1)OC)[N+](=O)[O-] |
| C9H10N2O4 | |
| MFCD00017018 | |
| n-4-methoxy-2-nitrophenyl acetamide, 4'-methoxy-2'-nitroacetanilide, 4-methoxy-2-nitroacetanilide, 2'-nitro-p-acetanisidide, acetamide, n-4-methoxy-2-nitrophenyl, 2-nitro-p-acetanisidide, p-acetanisidide, 2-nitro, 4-acetamido-3-nitroanisole, 2-nitro-4-methoxyacetanilide, p-acetanisidide, 2'-nitro | |
| N-(4-methoxy-2-nitrophenyl)acetamide |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 204-353-3
RTECSNumber : AE8310000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only