accessibility menu, dialog, popup

UserName

CAS RN 119-81-3

IUPAC Name: N-(4-methoxy-2-nitrophenyl)acetamide
Synonyms: 2-nitro-4-methoxyacetanilide
Molecular Weight (g/mol): 210.19
Molecular Formula: C9H10N2O4
InChi Key: QGEGALJODPBPGR-UHFFFAOYSA-N
SMILES: COC1=CC=C(NC(C)=O)C(=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results