missing translation for 'onlineSavingsMsg'
Learn More

4'-Methoxy-2'-nitroacetanilide, 98+%

Catalog No. AAL0421806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0421806 5 g
AAL0421814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0421806 Supplier Thermo Scientific Chemicals Supplier No. L0421806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 119-81-3 | C9H10N2O4 | 210.189 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 119-81-3
Molecular Formula C9H10N2O4
Molecular Weight (g/mol) 210.189
MDL Number MFCD00017018
InChI Key QGEGALJODPBPGR-UHFFFAOYSA-N
Synonym n-4-methoxy-2-nitrophenyl acetamide, 4'-methoxy-2'-nitroacetanilide, 4-methoxy-2-nitroacetanilide, 2'-nitro-p-acetanisidide, acetamide, n-4-methoxy-2-nitrophenyl, 2-nitro-p-acetanisidide, p-acetanisidide, 2-nitro, 4-acetamido-3-nitroanisole, 2-nitro-4-methoxyacetanilide, p-acetanisidide, 2'-nitro
PubChem CID 67075
IUPAC Name N-(4-methoxy-2-nitrophenyl)acetamide
SMILES CC(=O)NC1=C(C=C(C=C1)OC)[N+](=O)[O-]

Specifications

Melting Point 117°C to 118°C
Quantity 5 g
Beilstein 2809678
Formula Weight 210.19
Percent Purity ≥98%
Chemical Name or Material 4'-Methoxy-2'-nitroacetanilide
Hazard Category H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 204-353-3
RTECSNumber AE8310000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only