missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxy-2,6-dimethylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$145.40 - $145.40
Specifications
| CAS | 361543-99-9 |
|---|---|
| Physical Form | Crystal-Powder at 20°C |
| Molecular Formula | C9H13BO3 |
| MDL Number | MFCD03452755 |
| InChI Key | UOSSBXMFWPYHEF-UHFFFAOYSA-N |
Specifications
| 361543-99-9 | |
| C9H13BO3 | |
| UOSSBXMFWPYHEF-UHFFFAOYSA-N | |
| (4-methoxy-2,6-dimethylphenyl)boronic acid | |
| 3716303 |
| Crystal-Powder at 20°C | |
| MFCD03452755 | |
| B(C1=C(C=C(C=C1C)OC)C)(O)O | |
| 180.01 | |
| 180.01 |