accessibility menu, dialog, popup

UserName

Check Out the New Spotlight Offers and Get Up to 50% Off

Save Today

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 361543-99-9

BH3COH3CCH3HOOH

CAS RN 361543-99-9

IUPAC Name: (4-methoxy-2,6-dimethylphenyl)boronic acid
Synonyms: 4-methoxy-2,6-dimethylphenylboronic acid
Molecular Weight (g/mol): 180.01
Molecular Formula: C9H13BO3
InChi Key: UOSSBXMFWPYHEF-UHFFFAOYSA-N
SMILES: COC1=CC(C)=C(B(O)O)C(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.