missing translation for 'onlineSavingsMsg'
Learn More

4-(Methanesulfonamido)phenylboronic acid, 97%

Catalog No. AC445470050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC445470050 5 g
AC445470010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC445470050 Supplier Thermo Scientific Chemicals Supplier No. 445470050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 380430-57-9 | C7H10BNO4S | 215.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 380430-57-9
Molecular Formula C7H10BNO4S
Molecular Weight (g/mol) 215.03
MDL Number MFCD02179473
InChI Key NDVJJEADFLTFCD-UHFFFAOYSA-N
Synonym 4-methylsulfonylamino phenylboronic acid, 4-methylsulfonamido phenylboronic acid, 4-methanesulfonylaminophenylboronic acid, 4-methylsulfonylaminophenylboronic acid, 4-methanesulfonylamino phenylboronic acid, 4-methylsulfonamido phenyl boronic acid, 4-methanesulfonamidophenyl boronic acid, 4-methanesulfonamido phenyl boronic acid, 4-methylsulphonyl amino benzeneboronic acid, 4-methylsulfonylamino benzeneboronic acid
PubChem CID 2773537
IUPAC Name [4-(methanesulfonamido)phenyl]boronic acid
SMILES CS(=O)(=O)NC1=CC=C(C=C1)B(O)O

Specifications

Melting Point 234°C to 236°C
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 215.04
Percent Purity 97%
Chemical Name or Material 4-(Methanesulfonamido)phenylboronic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only