accessibility menu, dialog, popup

UserName

CAS RN 380430-57-9

IUPAC Name: (4-methanesulfonamidophenyl)boronic acid
Synonyms: 4-methanesulfonamidophenylboronic acid
Molecular Weight (g/mol): 215.03
Molecular Formula: C7H10BNO4S
InChi Key: NDVJJEADFLTFCD-UHFFFAOYSA-N
SMILES: CS(=O)(=O)NC1=CC=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results