missing translation for 'onlineSavingsMsg'
Learn More

4-Isothiocyanato-2-(trifluoromethyl)benzonitrile 98.0+%, TCI America™

Catalog No. I09871G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I09871G 1 g
I09875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I09871G Supplier TCI America Supplier No. I09871G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 143782-23-4
Molecular Formula C9H3F3N2S
Molecular Weight (g/mol) 228.192
MDL Number MFCD09800709
InChI Key TYXKOMAQTWRDCR-UHFFFAOYSA-N
Synonym 4-Cyano-3-(trifluoromethyl)phenyl Isothiocyanate, Isothiocyanic Acid 4-Cyano-3-(trifluoromethyl)phenyl Ester
PubChem CID 11615540
IUPAC Name 4-isothiocyanato-2-(trifluoromethyl)benzonitrile
SMILES C1=CC(=C(C=C1N=C=S)C(F)(F)F)C#N

Specifications

Melting Point 41°C
Color Yellow
Quantity 1 g
Formula Weight 228.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Isothiocyanato-2-(trifluoromethyl)benzonitrile