accessibility menu, dialog, popup

UserName

CAS RN 143782-23-4

IUPAC Name: 4-isothiocyanato-2-(trifluoromethyl)benzonitrile
Synonyms: 4-isothiocyanato-2-(trifluoromethyl)benzonitrile
Molecular Weight (g/mol): 228.19
Molecular Formula: C9H3F3N2S
InChi Key: TYXKOMAQTWRDCR-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=C(C=CC(=C1)N=C=S)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results