missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylphenoxyacetic acid, 98+%

Catalog No. AAL0110114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0110114 25 g
AAL0110106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0110114 Supplier Thermo Scientific Chemicals Supplier No. L0110114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1643-16-9 | C11H14O3 | 194.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1643-16-9
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00014364
InChI Key FPVCSFOUVDLTDG-UHFFFAOYSA-N
Synonym 4-isopropylphenoxyacetic acid, 2-4-isopropylphenoxy acetic acid, 4-isopropylphenoxy acetic acid, p-i-propylphenoxyacetic acid, 4-isopropyl-phenoxy-acetic acid, 4-propan-2-yl phenoxy acetic acid, 2-4-methylethyl phenoxy acetic acid, 4-isopropylphenoxyaceticacid, akos bbb/199, 2-4-propan-2-ylphenoxy acetic acid
PubChem CID 137131
IUPAC Name 2-(4-propan-2-ylphenoxy)acetic acid
SMILES CC(C)C1=CC=C(OCC(O)=O)C=C1

Specifications

Melting Point 85°C to 87°C
Quantity 25 g
Beilstein 2261017
Formula Weight 194.23
Percent Purity ≥98%
Chemical Name or Material 4-Isopropylphenoxyacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only