missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylbenzenesulfonamide, 97%

Catalog No. AAH3295003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3295003 1 g
AAH3295006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3295003 Supplier Thermo Scientific Chemicals Supplier No. H3295003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6335-39-3 | C9H13NO2S | 199.268 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6335-39-3
Molecular Formula C9H13NO2S
Molecular Weight (g/mol) 199.268
MDL Number MFCD00457071
InChI Key WVOWEROKBOQYLJ-UHFFFAOYSA-N
Synonym 4-isopropylbenzenesulfonamide, p-cumenesulfonamide, 4-1-methylethyl benzenesulfonamide, 4-propan-2-yl benzene-1-sulfonamide, 4-i-propyl benzenesulfonamide, 4-methylethyl benzenesulfonamide, p-isopropylbenzenesulfonamide, 4-isopropylbenzenesulphonamide, 4-iso-propylbenzenesulfonamide, 4-isopropyl-benzenesulfonamide
PubChem CID 232314
IUPAC Name 4-propan-2-ylbenzenesulfonamide
SMILES CC(C)C1=CC=C(C=C1)S(=O)(=O)N

Specifications

Melting Point 103°C to 106°C
Quantity 1 g
Formula Weight 199.27
Percent Purity 97%
Chemical Name or Material 4-Isopropylbenzenesulfonamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only